Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allantoin 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-(2,5-Dioxoimidazolidin-4-yl)urea

A15571-30 A15571-0B A15571-30 A15571-0I A15571-0I A15571-0B
AAA15571-0IEA 599.99 USD InStock
CAAAA15571-0I AAA15571-0I CAAAA15571-0B AAA15571-30 CAAAA15571-30 AAA15571-0B
Allantoin 98%
Allantoin


Formula: C₄H₆N₄O₃
MW: 158.12
Melting Pt: ca 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005260
CAS Number: 97-59-6
EINECS: 202-592-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty