Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pentafluorophenol 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2,3,4,5,6-Pentafluorophenol

A15574-14 A15574-22 A15574-06 A15574-06 A15574-14 A15574-22
AAA15574-14EA 140.99 USD InStock
CAAAA15574-22 CAAAA15574-14 AAA15574-14 CAAAA15574-06 AAA15574-22 AAA15574-06
Pentafluorophenol 99%
Pentafluorophenol


Formula: C₆HF₅O
MW: 184.07 g/mol
Boiling Pt: 143 °C
Melting Pt: 32…36 °C
Density: 1.757
Flash Pt: 72 °C(161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002156
CAS Number: 771-61-9
EINECS: 212-235-8
UN: 2923
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty