Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromooctadecane 97%

Supplier: Thermo Scientific Chemicals

A15588-22 A15588-0E A15588-36 A15588-0E A15588-36 A15588-22
AAA15588-22EA 30.22 USD InStock
CAAAA15588-36 CAAAA15588-0E AAA15588-22 AAA15588-0E AAA15588-36 CAAAA15588-22
1-Bromooctadecane 97%
1-Bromooctadecane


Formula: BrH₂C(CH₂)₁₆CH₃
MW: 333.4 g/mol
Boiling Pt: 150…152 °C/0.1 mm
Melting Pt: 25…30 °C
Density: 0.976
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000231
CAS Number: 112-89-0
EINECS: 204-013-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty