Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromophenylhydrazinium chloride 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Bromophenylhydrazine hydrochloride 1-(4-Bromophenyl)hydrazine hydrochloride

A15590-06 A15590-14 A15590-06 A15590-14
AAA15590-06EA 71.19 USD InStock
CAAAA15590-14 CAAAA15590-06 AAA15590-06 AAA15590-14
4-Bromophenylhydrazinium chloride 97%
4-Bromophenylhydrazinium chloride


Formula: C₆H₈BrClN₂
MW: 223.50 g/mol
Melting Pt: ca 215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012941
CAS Number: 622-88-8
EINECS: 255-590-4
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty