Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-n-Nonylphenol 98+%

Supplier: Thermo Scientific Chemicals
Danger

A15609-06 A15609-03
AAA15609-06EA 226 USD InStock
AAA15609-06 AAA15609-03
4-n-Nonylphenol 98+%
4-n-Nonylphenol


Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Boiling Pt: 127…130 °C/27 mm
Melting Pt: 43…46 °C
Density: 0.937
Flash Pt: 150 °C(302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002396
CAS Number: 104-40-5
EINECS: 203-199-4
UN: 2430
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty