Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-tert-Butylcyclohexanone 99%

Supplier: Thermo Scientific Chemicals
Warning

A15612-22 A15612-36 A15612-36 A15612-22
AAA15612-22EA 67.19 USD InStock
CAAAA15612-36 AAA15612-22 AAA15612-36 CAAAA15612-22
4-tert-Butylcyclohexanone 99%
4-tert-Butylcyclohexanone


Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 113…116 °C/20 mm
Melting Pt: 47…51 °C
Density: 0.893
Flash Pt: 96 °C(204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001642
CAS Number: 98-53-3
EINECS: 202-678-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty