Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl malonyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ethyl Malonyl Chloride

A15616-06 A15616-22 A15616-22 A15616-06 A15616-14 A15616-14
AAA15616-06EA 68 USD InStock
AAA15616-06 AAA15616-14 AAA15616-22 CAAAA15616-22 CAAAA15616-06 CAAAA15616-14
Ethyl malonyl chloride 95%
Ethyl malonyl chloride


Formula: C₅H₇ClO₃
MW: 150.56 g/mol
Boiling Pt: 83 °C/22 mm
Density: 1.189
Flash Pt: 67 °C(152 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000736
CAS Number: 36239-09-5
EINECS: 252-934-5
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty