Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloro(dimethyl)(phenyl)silane 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Chlorodimethylphenylsilane

A15638-22 A15638-14 A15638-06 A15638-22 A15638-14 A15638-06
AAA15638-14EA 138.99 USD InStock
CAAAA15638-06 AAA15638-14 CAAAA15638-14 AAA15638-06 CAAAA15638-22 AAA15638-22
Chloro(dimethyl)(phenyl)silane 97+%
Chloro(dimethyl)(phenyl)silane


Formula: C₈H₁₁ClSi
MW: 170.71 g/mol
Boiling Pt: 192…193 °C
Density: 1.032
Flash Pt: 72 °C(161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000499
CAS Number: 768-33-2
EINECS: 212-193-0
UN: 2987
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty