Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-1,4-dimethoxybenzene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Bromo-2,5-dimethoxybenzene

A15640-18 A15640-30 A15640-09
AAA15640-18EA 213.99 USD InStock
AAA15640-18 AAA15640-30 AAA15640-09
2-Bromo-1,4-dimethoxybenzene 98%
2-Bromo-1,4-dimethoxybenzene


Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Boiling Pt: 260…262 °C
Density: 1,441
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008355
CAS Number: 25245-34-5
EINECS: 246-756-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty