Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-4-chloroacetoacetate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ethyl 4-chloro-3-oxobutanoate

A15641-0B A15641-30 A15641-0B A15641-18 A15641-18 A15641-30
AAA15641-18EA 34.79 USD InStock
CAAAA15641-18 AAA15641-18 CAAAA15641-30 AAA15641-0B AAA15641-30 CAAAA15641-0B
Ethyl-4-chloroacetoacetate 97%
Ethyl 4-chloroacetoacetate


Formula: C₆H₉ClO₃
MW: 164.59 g/mol
Boiling Pt: 220 °C
Melting Pt: –8 °C
Density: 1.212
Flash Pt: 92 °C(198 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000939
CAS Number: 638-07-3
EINECS: 211-317-0
UN: 2927
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty