Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Methyl)triphenylphosphonium iodide 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methyltriphenylphosphonium iodide

A15644-22 A15644-14 A15644-22 A15644-36 A15644-14 A15644-36
AAA15644-22EA 179.99 USD InStock
CAAAA15644-22 AAA15644-22 CAAAA15644-14 CAAAA15644-36 AAA15644-14 AAA15644-36
(Methyl)triphenylphosphonium iodide 98%
(Methyl)triphenylphosphonium iodide


Formula: C₁₉H₁₈IP
MW: 404.23 g/mol
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00066175
CAS Number: 2065-66-9
EINECS: 218-178-5
UN: 3464
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty