Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzotrichloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (Trichloromethyl)benzene α,α,α-Trichlorotoluene Phenylchloroform

A15652-36 A15652-36 A15652-0E A15652-0E
AAA15652-36EA 179.99 USD InStock
AAA15652-36 AAA15652-0E CAAAA15652-36 CAAAA15652-0E
Benzotrichloride 98%
Benzotrichloride


Formula: C₇H₅Cl₃
MW: 195.47 g/mol
Boiling Pt: 90…91 °C/15 mm
Melting Pt: –8 to –.5 °C
Density: 1.38
Flash Pt: 97 °C(207 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000786
CAS Number: 98-07-7
EINECS: 202-634-5
UN: 2226
ADR: "8",II
Merck Index: 13,01110
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/33/763133.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty