Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorophenylboronic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Chlorobenzeneboronic acid Dihydroxy(4-chlorophenyl)borane (4-Chlorophenyl)metaboric acid

A15657-14 A15657-06 A15657-03 A15657-14 A15657-06 A15657-03
AAA15657-06EA 61.59 USD InStock
CAAAA15657-14 CAAAA15657-03 AAA15657-06 AAA15657-14 AAA15657-03 CAAAA15657-06
4-Chlorophenylboronic acid 98+%
4-Chlorophenylboronic acid


Formula: C₆H₆BClO₂
MW: 156.38 g/mol
Melting Pt: 268…277 °C
Density: ∼1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039137
CAS Number: 1679-18-1
EINECS: 216-845-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty