Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-α-Phenylglycine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: H-D-Phg-OHD-(-)-2-Phenylglycine(R)-(-)-α-Aminophenylacetic acid

A15669-14 A15669-14 A15669-22 A15669-36 A15669-22 A15669-36
AAA15669-14EA 29.39 USD InStock
CAAAA15669-22 CAAAA15669-14 CAAAA15669-36 AAA15669-14 AAA15669-36 AAA15669-22
D-(-)-α-Phenylglycine 99%
D(-)-α-Phenylglycine


Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 277 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008061
CAS Number: 875-74-1
EINECS: 212-876-3
Order Now
Specification Test Results
Optical rotation -154° (c=1 in 1N HCl)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty