Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methyl-2-cyclohexen-1-one 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Methylcyclohex-2-enone

A15704-22 A15704-14 A15704-09 A15704-18 A15704-22 A15704-18 A15704-14 A15704-09
AAAA15704-18EA 260.99 USD InStock
CAAAAA15704-09 AAAA15704-18 CAAAA15704-22 AAAA15704-09 CAAAA15704-14 AAA15704-22 CAAAAA15704-18 AAA15704-14
3-Methyl-2-cyclohexen-1-one 98%
3-Methyl-2-cyclohexen-1-one


Formula: C₇H₁₀O
MW: 110.16 g/mol
Boiling Pt: 199…200 °C
Melting Pt: 198…199 °C
Density: 0.971
Flash Pt: 68 °C(154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001581
CAS Number: 1193-18-6
EINECS: 214-769-7
ADR: 3,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty