Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Benzyloxy)benzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A15715-03 A15715-06 A15715-03 A15715-14 A15715-06 A15715-14
AAA15715-03EA 24.99 USD InStock
CAAAA15715-06 CAAAA15715-03 AAA15715-03 AAA15715-14 CAAAA15715-14 AAA15715-06
4-(Benzyloxy)benzoic acid 98%
4-(Benzyloxy)benzoic acid


Formula: C₁₄H₁₂O₃
Melting Pt: 187…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00016527
CAS Number: 1486-51-7
EINECS: 216-066-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty