Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dextrin, precipitated by alcohol

Supplier: Thermo Scientific Chemicals

A15717-22 A15717-36 A15717-22 A15717-36
AAA15717-22EA 47 USD InStock
CAAAA15717-22 AAA15717-22 AAA15717-36 CAAAA15717-36
Dextrin, precipitated by alcohol
Dextrin


Formula: (C₆H₁₂O₆)n
MW: 180.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081554
CAS Number: 9004-53-9
EINECS: 232-675-4
Merck Index: 13,02971
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty