Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Chloro-4-nitrotoluene 99%

Supplier: Thermo Scientific Chemicals
Danger

A15749-30 A15749-09 A15749-18 A15749-30 A15749-18 A15749-09
AAA15749-18EA 77.69 USD InStock
AAA15749-18 AAA15749-09 CAAAA15749-09 CAAAA15749-30 CAAAA15749-18 AAA15749-30
α-Chloro-4-nitrotoluene 99%
α-Chloro-4-nitrotoluene


Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00007374
CAS Number: 100-14-1
EINECS: 202-822-7
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty