Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Hydroxynicotinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Hydroxy-3-pyridinecarboxylic acid 6-Hydroxypyridine-3-carboxylic acid

A15802-22 A15802-06 A15802-14 A15802-14 A15802-06 A15802-22
AAA15802-06EA 26.39 USD InStock
CAAAA15802-14 CAAAA15802-22 AAA15802-06 CAAAA15802-06 AAA15802-14 AAA15802-22
6-Hydroxynicotinic acid 98%
6-Hydroxynicotinic acid
Metabolite of nicotinamide. Stimulates adrenocortical secretion


Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 314…316 °C
Storage Temperature: Ambient
MDL Number: MFCD00006277
CAS Number: 5006-66-6
EINECS: 225-682-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty