Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Epichlorhydrin 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (±)-1-Chloro-2,3-epoxypropane (±)-3-Chloro-1,2-epoxypropane 1-Chloro-2,3-epoxypropane 3-Chloro-1,2-epoxypropane Epichlorhydrin (±)-Glycerol epichlorhydrin

A15823-36 A15823-22 A15823-0E A15823-36 A15823-0E A15823-22
AAA15823-22EA 39.32 USD InStock
CAAAA15823-22 CAAAA15823-36 CAAAA15823-0E AAA15823-22 AAA15823-0E AAA15823-36
(±)-Epichlorhydrin 99%
(±)-Epichlorhydrin
A cyclodextrin cross-linking reagent


Formula: C₃H₅ClO
MW: 92.52 g/mol
Boiling Pt: 115…117 °C
Melting Pt: –57 °C
Density: 1.182
Flash Pt: 32 °C(90 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005132
CAS Number: 106-89-8
EINECS: 203-439-8
UN: 2023
ADR: "6.1",II
Merck Index: 13,03642
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty