Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Glucurono-6,3-lactone 99%

Supplier: Thermo Scientific Chemicals
Synonyms: D-(+)-Glucuronolactone(R)-2-((2S,3R,4S)-3,4-Dihydroxy-5-oxotetrahydrofuran-2-yl)-2-hydroxyacetaldehyde

A15861-36 A15861-22 A15861-36 A15861-14 A15861-14 A15861-22
AAA15861-22EA 73.09 USD InStock
CAAAA15861-22 CAAAA15861-36 CAAAA15861-14 AAA15861-22 AAA15861-36 AAA15861-14
D-(+)-Glucurono-6,3-lactone 99%
D(+)-Glucurono-6,3-lactone


Formula: C₆H₈O₆
MW: 176.12 g/mol
Melting Pt: 174 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00135622
CAS Number: 32449-92-6
EINECS: 251-053-3
Order Now
Specification Test Results
Optical rotation +19° (c=8 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty