Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-tert-Butylphenol 99%

Supplier: Thermo Scientific Chemicals
Danger

A15871-36 A15871-36 A15871-0E A15871-0E
AAA15871-36EA 40.26 USD InStock
CAAAA15871-0E AAA15871-36 CAAAA15871-36 AAA15871-0E
4-tert-Butylphenol 99%
4-tert-Butylphenol


Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 236…238 °C
Melting Pt: 97…101 °C
Density: 0.908
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002367
CAS Number: 98-54-4
EINECS: 202-679-0
UN: 3077
ADR: "9",III
Merck Index: 13,01584
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty