Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-fluorotoluene 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Bromo-3-fluoro-4-methylbenzene

A15891-22 A15891-14 A15891-06 A15891-14 A15891-06 A15891-22
AAA15891-14EA 115.99 USD InStock
CAAAA15891-06 AAA15891-14 AAA15891-22 AAA15891-06 CAAAA15891-14 CAAAA15891-22
4-Bromo-2-fluorotoluene 99%
4-Bromo-2-fluorotoluene


Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 66…68 °C/8 mm
Density: 1,492
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013551
CAS Number: 51436-99-8
EINECS: 257-201-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty