Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Valine methyl ester hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Val-OMe·HClL-2-Aminoisovaleric Acid Methyl Ester HydrochlorideL-Methyl valinate hydrochloride

A15892-18 A15892-18 A15892-09 A15892-09
AAA15892-18EA 214.99 USD InStock
CAAAA15892-18 CAAAA15892-09 AAA15892-18 AAA15892-09
L-Valine methyl ester hydrochloride 99%
L-Valine methyl ester hydrochloride


Formula: C₆H₁₄ClNO₂
MW: 167.63 g/mol
Melting Pt: 166 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012497
CAS Number: 6306-52-1
EINECS: 228-620-9
Order Now
Specification Test Results
Optical rotation +16° (c=2 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty