Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Cyanobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Formylbenzonitrile

A15943-03 A15943-03 A15943-14 A15943-06 A15943-06 A15943-14
AAA15943-06EA 94.39 USD InStock
AAA15943-06 CAAAA15943-03 CAAAA15943-14 AAA15943-03 AAA15943-14 CAAAA15943-06
3-Cyanobenzaldehyde 97%
3-Cyanobenzaldehyde


Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 209…210 °C
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00003344
CAS Number: 24964-64-5
EINECS: 246-549-1
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty