Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Caffeic acid predominantly trans 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3,4-Dihydroxycinnamic acid3-(3,4-Dihydroxyphenyl)acrylic acid

A15950-18 A15950-09 A15950-18 A15950-09
AAA15950-18EA 261.99 USD InStock
CAAAA15950-18 AAA15950-18 AAA15950-09 CAAAA15950-09
Caffeic acid predominantly trans 98+%
Caffeic acid
Inhibitor of ornithine decarboxylase and protein tyrosine kinase


Formula: C₉H₈O₄
MW: 180.16 g/mol
Melting Pt: ca 202 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004392
CAS Number: 331-39-5
EINECS: 206-361-2
Merck Index: 12,01673
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty