Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Methionine methyl ester hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Met-OMe.HClH-Met-OMe·HCl

A15952-06 A15952-22 A15952-06 A15952-14 A15952-22 A15952-14
AAA15952-06EA 36.09 USD InStock
CAAAA15952-06 AAA15952-06 CAAAA15952-14 CAAAA15952-22 AAA15952-22 AAA15952-14
L-Methionine methyl ester hydrochloride 99%
L-Methionine methyl ester hydrochloride


Formula: C₆H₁₄ClNO₂S
MW: 199.7 g/mol
Melting Pt: 151…152 °C
Storage Temperature: Freezer
MDL Number: MFCD00012491
CAS Number: 2491-18-1
EINECS: 219-651-9
Order Now
Specification Test Results
Optical rotation +26° (c=5 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty