Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methylbenzimidazole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Methyl-1H-benzimidazole

A15957-06 A15957-22 A15957-22 A15957-14 A15957-14 A15957-06
AAA15957-22EA 597.99 USD InStock
CAAAA15957-22 AAA15957-22 CAAAA15957-14 CAAAA15957-06 AAA15957-14 AAA15957-06
5-Methylbenzimidazole 98%
5-Methylbenzimidazole


Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 169…172 °C/1 mm
Melting Pt: 112…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00010740
CAS Number: 614-97-1
EINECS: 210-401-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty