Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Methoxy-1-tetralone 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 6-Methoxy-3,4-dihydronaphthalen-1(2H)-one6-Methoxy-1,2,3,4-tetrahydro-1-naphthalenone

A15967-14 A15967-22 A15967-14 A15967-06 A15967-06 A15967-22
AAA15967-14EA 56.19 USD InStock
CAAAA15967-06 AAA15967-14 CAAAA15967-14 AAA15967-22 AAA15967-06 CAAAA15967-22
6-Methoxy-1-tetralone 99%
6-Methoxy-1-tetralone


Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Boiling Pt: 171 °C/11 mm
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00001695
CAS Number: 1078-19-9
EINECS: 214-078-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty