Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl cinnamate predominantly trans 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Methyl 3-phenylpropenoateMethyl 3-phenyl-2-propenoate

A15975-36 A15975-36 A15975-22 A15975-0E A15975-22 A15975-0E
AAA15975-36EA 105.99 USD InStock
AAA15975-36 CAAAA15975-36 AAA15975-22 CAAAA15975-0E CAAAA15975-22 AAA15975-0E
Methyl cinnamate predominantly trans 99%
Methyl cinnamate


Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 262…264 °C
Melting Pt: 34…37 °C
Density: 1.042
Flash Pt: 123 °C(253 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008458
CAS Number: 103-26-4
EINECS: 203-093-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty