Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-5-(trifluoromethyl)aniline 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Chloro-α,α,α-trifluoro-m-toluidine 3-Amino-4-chloro-α,α,α-trifluorotoluene 3-Amino-4-chloro-α,α,α-trifluortoluene

A16004-36 A16004-22 A16004-22 A16004-36
AAA16004-36EA 168.79 USD InStock
AAA16004-36 CAAAA16004-36 AAA16004-22 CAAAA16004-22
2-Chloro-5-(trifluoromethyl)aniline 97%
2-Chloro-5-(trifluoromethyl)aniline


Formula: C₇H₅ClF₃N
MW: 195.57 g/mol
Boiling Pt: 82…83 °C/9 mm
Melting Pt: 8…10 °C
Density: 1.424
Flash Pt: 75 °C(167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007673
CAS Number: 121-50-6
EINECS: 204-475-7
UN: 3082
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty