Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrazolium violet 98+%

Supplier: Thermo Scientific Chemicals
Warning

A16008-06 A16008-14 A16008-03 A16008-03 A16008-14 A16008-06
AAA16008-03EA 46.09 USD InStock
AAA16008-03 AAA16008-14 AAA16008-06 CAAAA16008-06 CAAAA16008-14 CAAAA16008-03
Tetrazolium violet 98+%
Tetrazolium Violet


Formula: C₂₃H₁₇ClN₄
Melting Pt: 247…249 °C
Storage Temperature: Ambient
MDL Number: MFCD00011875
CAS Number: 1719-71-7
EINECS: 217-008-7
UN: 1325
ADR: "4.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty