Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Ethylphthalimide 98%

Supplier: Thermo Scientific Chemicals
Warning

A16014-14 A16014-06 A16014-14 A16014-22 A16014-06 A16014-22
AAAA16014-22EA 1078.96 USD InStock
AAAA16014-22 AAA16014-06 CAAAA16014-06 AAA16014-14 CAAAAA16014-22 CAAAA16014-14
N-Ethylphthalimide 98%
N-Ethylphthalimide


Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00014583
CAS Number: 5022-29-7
EINECS: 225-707-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty