Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-ɑ-Boc-L-asparagine ≥98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Boc-Asn-OHBoc-L-Asparagine

A16019-06 A16019-06 A16019-14 A16019-14
AAA16019-06EA 26.99 USD InStock
AAA16019-06 AAA16019-14 CAAAA16019-14 CAAAA16019-06
N-ɑ-Boc-L-asparagine ≥98%
N-alpha-Boc-L-asparagine


Formula: C9H16N2O5
MW: 232.24 g/mol
Melting Pt: ca 170 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00038152
CAS Number: 7536-55-2
EINECS: 231-405-2
Order Now
Specification Test Results
Optical rotation -7.3° (c=2 in DMF)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty