Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Indigo carmine

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Acid Blue 74 Food blue No. 2 FD and C Blue No. 2

A16052-14 A16052-22 A16052-22 A16052-14
AAA16052-14EA 54 USD InStock
AAA16052-14 CAAAA16052-22 CAAAA16052-14 AAA16052-22
Indigo carmine
Indigo carmine
Useful in the staining of Negri bodies


Formula: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005723
CAS Number: 860-22-0
EINECS: 212-728-8
Merck Index: 13,04969
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty