Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glyoxylic acid monohydrate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Formylformic acid monohydrateOxoacetic acid monohydrate2-Oxoethanoic acid monohydrate

A16058-22 A16058-14 A16058-14 A16058-22
AAA16058-14EA 51.21 USD InStock
CAAAA16058-14 CAAAA16058-22 AAA16058-14 AAA16058-22
Glyoxylic acid monohydrate 97%
Glyoxylic acid monohydrate


Formula: C₂H₂O₃·1H₂O
MW: 92.05 g/mol
Boiling Pt: 100 °C
Melting Pt: 48…52 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00149497
CAS Number: 563-96-2
EINECS: 206-058-5
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty