Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(Trimethylsiloxy)cyclohexene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (Cyclohex-1-en-1-yloxy)trimethylsilane

A16099-09 A16099-30 A16099-18 A16099-09 A16099-30 A16099-18
AAA16099-30EA 1051.53 USD InStock
CAAAA16099-18 CAAAA16099-09 AAA16099-30 AAA16099-09 AAA16099-18 CAAAA16099-30
1-(Trimethylsiloxy)cyclohexene 98%
1-(Trimethylsiloxy)cyclohexene


Formula: C₉H₁₈OSi
MW: 170.33 g/mol
Boiling Pt: 165 °C
Density: 0.883
Flash Pt: 42 °C(107 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001541
CAS Number: 6651-36-1
EINECS: 229-675-1
ADR: 3,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty