Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluorotoluene 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-Fluoro-3-methylbenzene

A16107-22 A16107-14 A16107-30 A16107-30 A16107-22 A16107-14
AAA16107-30EA 90.49 USD InStock
CAAAAA16107-22 CAAAAA16107-14 AAA16107-30 CAAAA16107-30 AAAA16107-14 AAAA16107-22
3-Fluorotoluene 98%
3-Fluorotoluene


Formula: C₇H₇F
MW: 110.13 g/mol
Boiling Pt: 114…116 °C
Melting Pt: –87 °C
Density: 0.993
Flash Pt: 9 °C(48 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000339
CAS Number: 352-70-5
EINECS: 206-524-8
UN: 2388
ADR: "3",II
Merck Index: 13,04207
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty