Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyethylene glycol 4,000

Supplier: Thermo Scientific Chemicals
Synonyms: PEG 4,000PEG 4000

A16151-0I A16151-30 A16151-0B A16151-0B A16151-0I A16151-30
AAA16151-30EA 62.11 USD InStock
CAAAA16151-30 AAA16151-30 CAAAA16151-0I AAA16151-0B CAAAA16151-0B AAA16151-0I
Polyethylene glycol 4,000
Polyethylene glycol 4,000


Formula: H(OCH₂CH₂)nOH
MW: 4000 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 58…62 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD01779612
CAS Number: 25322-68-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty