Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-Dibenzoyl-L-tartaric acid monohydrate 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (2R,3R)-(-)-Di-O-benzoyl-tartaric acid monohydrate

A16180-22 A16180-0E A16180-0E A16180-36 A16180-36 A16180-22
AAA16180-22EA 51.29 USD InStock
AAA16180-22 CAAAA16180-0E AAA16180-0E AAA16180-36 CAAAA16180-22 CAAAA16180-36
(-)-Dibenzoyl-L-tartaric acid monohydrate 98+%
(-)-Dibenzoyl-L-tartaric acid monohydrate


Formula: C₁₈H₁₄O₈·1H₂O
MW: 376.33 (358.31 anhy)
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00149119
CAS Number: 62708-56-9
EINECS: 613-072-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty