Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Acetylindole 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Indol-3-yl methyl ketone 1-(3-Indolyl)-1-ethanone

A16221-06 A16221-14 A16221-22 A16221-06 A16221-14 A16221-22
AAA16221-06EA 71.79 USD InStock
AAA16221-06 CAAAA16221-22 CAAAA16221-14 AAA16221-22 CAAAA16221-06 AAA16221-14
3-Acetylindole 98%
3-Acetylindole


Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 144…145 °C (10 mmHg)
Melting Pt: 186…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00005626
CAS Number: 703-80-0
EINECS: 211-875-5
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty