Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Arginine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-(-)-Arginine hydrochloride H-D-Arg-OH.HCl

A16222-03 A16222-14 A16222-06 A16222-14 A16222-06 A16222-03
AAA16222-06EA 142.99 USD InStock
CAAAA16222-14 CAAAA16222-03 CAAAA16222-06 AAA16222-06 AAA16222-03 AAA16222-14
D-(-)-Arginine hydrochloride 99%
D(-)-Arginine hydrochloride


Formula: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
Melting Pt: 216…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00012620
CAS Number: 627-75-8
EINECS: 211-010-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty