Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Maltose monohydrate 95%

Supplier: Thermo Scientific Chemicals
Synonyms: beta-Maltose monohydrateMaltobiose hydrateMalt Sugar4-O-α-D-Glucopyranosyl-D-glucose hydrateMaltose monohydrate(2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate

A16266-36 A16266-22 A16266-36 A16266-22
AAA16266-36EA 165.51 USD InStock
AAA16266-36 AAA16266-22 CAAAA16266-22 CAAAA16266-36
D-(+)-Maltose monohydrate 95%
D(+)-Maltose monohydrate


Formula: C₁₂H₂₂O₁₁·1H₂O
MW: 360.32 g/mol
Melting Pt: ca 120 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149343
CAS Number: 6363-53-7
EINECS: 200-716-5
Merck Index: 13,05736
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/24/2302124.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty