Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Dichlorobenzene 99+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: p-Dichlorobenzene

A16290-36 A16290-0E A16290-22
AAAA16290-22EA 37.63 USD InStock
AAAA16290-22 AAA16290-0E
1,4-Dichlorobenzene 99+%
1,4-Dichlorobenzene


Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 173…174 °C
Melting Pt: 52…56 °C
Density: 1.248
Flash Pt: 65 °C(149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000604
CAS Number: 106-46-7
EINECS: 203-400-5
UN: 3077
ADR: "9",III
Merck Index: 13,03082
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty