Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibutyl ether 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Di-n-butyl ether

A16305-0F A16305-0F A16305-AP A16305-AP
AAA16305-0FEA 93 USD InStock
AAA16305-0F CAAAA16305-AP CAAAA16305-0F AAA16305-AP
Dibutyl ether 99%
Dibutyl ether


Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 141…143 °C
Melting Pt: –95 °C
Density: 0.764
Flash Pt: 25 °C(77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009461
CAS Number: 142-96-1
EINECS: 205-575-3
UN: 1149
ADR: "3",III
Merck Index: 13,01568
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty