Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+)-Dibenzoyl-D-tartaric acid monohydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

A16503-14 A16503-22 A16503-14 A16503-22
AAA16503-14EA 53.69 USD InStock
AAA16503-14 CAAAA16503-14 AAA16503-22 CAAAA16503-22
(+)-Dibenzoyl-D-tartaric acid monohydrate 99%
(+)-Dibenzoyl-D-tartaric acid monohydrate


Formula: C₁₈H₁₄O₈·1H₂O
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00150722
CAS Number: 80822-15-7
EINECS: 241-097-1
Order Now
Specification Test Results
Optical rotation +109° (c=2 in ethanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty