Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Hydroxy-o-tolyl-β-D-glucopyranoside 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: D(−)-Salicin Salicoside 2-(Hydroxymethyl)phenyl-β-D-glucopyranoside (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol

A16524-06 A16524-14 A16524-22 A16524-22 A16524-14 A16524-06
AAA16524-14EA 129.84 USD InStock
CAAAA16524-14 AAA16524-14 CAAAA16524-06 AAA16524-22 AAA16524-06 CAAAA16524-22
α-Hydroxy-o-tolyl-β-D-glucopyranoside 99%
α-Hydroxy-o-tolyl β-D-glucopyranoside
An analgesic and antipyretic. Substrate for beta-glucosidase


Formula: C₁₃H₁₈O₇
MW: 286.28 g/mol
Melting Pt: 200…203 °C
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006590
CAS Number: 138-52-3
EINECS: 205-331-6
Merck Index: 12,08476
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty