Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methyl-2-pentenoic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

A16587-18 A16587-18 A16587-30 A16587-30 A16587-09 A16587-09
AAA16587-18EA 128.99 USD InStock
AAA16587-18 CAAAA16587-30 AAA16587-09 CAAAA16587-09 AAA16587-30 CAAAA16587-18
2-Methyl-2-pentenoic acid 99%
2-Methyl-2-pentenoic acid


Formula: C₆H₁₀O₂
Boiling Pt: 120 °C/20 mm
Density: 0.978
Flash Pt: 107 °C(225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002655
CAS Number: 3142-72-1
EINECS: 221-552-0
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty