Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oleic acid tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Synonyms: (9Z)-9-Octadecenoic acid

A16663-0J A16663-AU A16663-0J A16663-AU
AAA16663-AUEA 82.39 USD InStock
CAAAA16663-0J AAA16663-AU CAAAA16663-AU AAA16663-0J
Oleic acid tech. 90%, Technical Grade
Oleic acid


Formula: C₁₈H₃₄O₂
MW: 282.47 g/mol
Boiling Pt: 286 °C/100 mm
Melting Pt: 13…14 °C
Density: 0.887
Storage Temperature: Refrigerator
MDL Number: MFCD00064242
CAS Number: 112-80-1
EINECS: 204-007-1
Merck Index: 13,06898
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/57/2298957.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty