Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Alanine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 3-Aminopropionic acid H-Bala-OH H-beta-Ala-OH

A16665-30 A16665-0B A16665-0I A16665-0I A16665-0B A16665-30
AAA16665-30EA 45.77 USD InStock
CAAAA16665-30 AAA16665-30 CAAAA16665-0I AAA16665-0B CAAAA16665-0B AAA16665-0I
β-Alanine 98%
β-Alanine


Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 201 °C (1013 hPa)
Melting Pt: ca 205 °C (dec.)
Density: 1.437
Storage Temperature: Ambient
MDL Number: MFCD00008200
CAS Number: 107-95-9
EINECS: 203-536-5
Merck Index: 13,00204
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty